accessibility menu, dialog, popup

UserName

CAS RN 2127-03-9

SSNN

CAS RN 2127-03-9

IUPAC Name: 2-(pyridin-2-yldisulfanyl)pyridine
Synonyms: 2,2'-dipyridyldisulfide
Molecular Weight (g/mol): 220.31
Molecular Formula: C10H8N2S2
InChi Key: HAXFWIACAGNFHA-UHFFFAOYSA-N
SMILES: S(SC1=CC=CC=N1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results