CAS RN 21553-46-8
IUPAC Name:
4-methoxy-3,5-dimethylbenzoate
Synonyms:
4-methoxy-3,5-dimethylbenzoate
Molecular Weight (g/mol):
179.2
Molecular Formula:
C10H11O3
InChi Key:
WXVQURJGDUNJCS-UHFFFAOYSA-M
SMILES:
COC1=C(C)C=C(C=C1C)C([O-])=O
1
–
1
of
1
results
1
3,5-Dimethyl-p-anisic acid, 98%, Thermo Scientific™
CAS: 21553-46-8 Molecular Formula: C10H12O3 Molecular Weight (g/mol): 180.2 MDL Number: MFCD00020309 InChI Key: WXVQURJGDUNJCS-UHFFFAOYSA-N Synonym: 3,5-dimethyl-4-methoxybenzoic acid,3,5-dimethyl-p-anisic acid,4-methoxy-3,5-dimethyl-benzoic acid,3,5-dimethyl anisic acid,benzoic acid, 4-methoxy-3,5-dimethyl,4-methoxy-3,5-dimethylbenzoicacid,acmc-209flo,ksc496g8f,3,5-dimethyl-4-methoxy-benzoic acid,4-methoxy-3,5-dimethyl benzoic acid PubChem CID: 88944 IUPAC Name: 4-methoxy-3,5-dimethylbenzoic acid SMILES: CC1=CC(=CC(=C1OC)C)C(=O)O