missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-p-anisic acid, 98%, Thermo Scientific™

Product Code. 10052290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10052290 5 g Glass bottle 5g
10646012 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10052290 Supplier Thermo Scientific Acros Supplier No. 187130050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 21553-46-8
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00020309
InChI Key WXVQURJGDUNJCS-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-methoxybenzoic acid, 3,5-dimethyl-p-anisic acid, 4-methoxy-3,5-dimethyl-benzoic acid, 3,5-dimethyl anisic acid, benzoic acid, 4-methoxy-3,5-dimethyl, 4-methoxy-3,5-dimethylbenzoicacid, acmc-209flo, ksc496g8f, 3,5-dimethyl-4-methoxy-benzoic acid, 4-methoxy-3,5-dimethyl benzoic acid
PubChem CID 88944
IUPAC Name 4-methoxy-3,5-dimethylbenzoic acid
SMILES CC1=CC(=CC(=C1OC)C)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 190°C to 194°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H2(CH3)2CO2H
Quantity 5 g
Formula Weight 180.2
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 3, 5-Dimethyl-p-anisic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dimethyl-p-anisic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.