accessibility menu, dialog, popup

UserName

CAS RN 217-59-4

IUPAC Name: triphenylene
Synonyms: triphenylene 9,10-benzophenanthrene 1,2,3,4-dibenznaphthalene 9,10-benzphenanthrene isochrysene benzo(l)phenanthrene
Molecular Weight (g/mol): 228.29
Molecular Formula: C18H12
InChi Key: SLGBZMMZGDRARJ-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results