accessibility menu, dialog, popup

UserName

CH3H3COHNOONHH3CCH3CH3NCH3CH3H3CCH3

CAS RN 217171-01-2

IUPAC Name: N-{2-[4-(benzyloxy)phenyl]-1-(tert-butylcarbamoyl)ethyl}-2-{[(4-tert-butylphenyl)methyl](methyl)amino}-4-methylpentanamide
Synonyms: N-{2-[4-(benzyloxy)phenyl]-1-(tert-butylcarbamoyl)ethyl}-2-{[(4-tert-butylphenyl)methyl](methyl)amino}-4-methylpentanamide
Molecular Weight (g/mol): 599.86
Molecular Formula: C38H53N3O3
InChi Key: GCDHMGROXQUFNR-UHFFFAOYNA-N
SMILES: CC(C)CC(N(C)CC1=CC=C(C=C1)C(C)(C)C)C(=O)NC(CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(=O)NC(C)(C)C
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results