missing translation for 'onlineSavingsMsg'
Learn More

PD173212, MedChemExpress

Product Code. 16443828
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443828 10 mg 10mg
16430086 2 mg 2mg
16406557 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16443828 Supplier Medchem Express Supplier No. HY103318

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PD173212 is a selective N-type voltage sensitive calcium channel (VSCC) blocker, with an IC50 of 36 nM in IMR-32 assays.

PD173212 is a selective N-type voltage sensitive calcium channel (VSCC) blocker, with an IC50 of 36 nM in IMR-32 assays.

Specifications

Chemical Name or Material PD173212
CAS 217171-01-2
Color White
Purity Grade Notes Research
Molecular Formula C38H53N3O3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (166.71 mM)
SMILES O=C(NC(C)(C)C)[C@@H](NC([C@H](CC(C)C)N(C)CC1=CC=C(C(C)(C)C)C=C1)=O)CC2=CC=C(OCC3=CC=CC=C3)C=C2
Molecular Weight (g/mol) 599.85
Formula Weight 599.85
Percent Purity 95.01%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.