accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 21905-56-6

IUPAC Name: methyl 2-phenoxybenzoate
Synonyms: methyl 2-phenoxybenzoate
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: PUGYLBSXMKBSRP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1OC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results