missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-phenoxybenzoate, 99%

Product Code. 11323606
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323606 50 g 50g
11313606 10 g 10g
11333606 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323606 Supplier Thermo Scientific Alfa Aesar Supplier No. L14584.18
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21905-56-6 | C14H12O3 | 228.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21905-56-6
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD03093064
InChI Key PUGYLBSXMKBSRP-UHFFFAOYSA-N
PubChem CID 607606
IUPAC Name methyl 2-phenoxybenzoate
SMILES COC(=O)C1=CC=CC=C1OC1=CC=CC=C1

Specifications

Boiling Point 130°C (0.5 mmHg)
Refractive Index 1.576
Quantity 50 g
Beilstein 2115719
Synonym 2-phenoxybenzoic acid methyl ester, benzoic acid, 2-phenoxy-, methyl ester, methyl o-phenoxybenzoate, acmc-1ce6i, #, benzoic acid,2-phenoxy-, methyl ester, benzoic acid, o-phenoxy-, methyl ester
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material Methyl 2-phenoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.