CAS RN 2196183-71-6
CAS RN 2196183-71-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Benzyl Benzydamine Hydrochloride, TRC
CAS: 2196183-71-6 Molecular Formula: C26 H29 N3 O . Cl H Molecular Weight (g/mol): 435.99 Synonym: 1-Propanamine, 3-[[1,5-bis(phenylmethyl)-1H-indazol-3-yl]oxy]-N,N-dimethyl-, hydrochloride (1:1),3-(1,5-Dibenzyl-1H-indazol-3-yl)oxypropyldimethylamine hydrochloride,5-Benzyl Benzydamine hydrochloride,5-Benzylbenzydamine hydrochloride IUPAC Name: 3-(1,5-dibenzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride SMILES: Cl.CN(C)CCCOc1nn(Cc2ccccc2)c3ccc(Cc4ccccc4)cc13