accessibility menu, dialog, popup

UserName

CH3H3CCH3OHOHOH3CH3CCH3(E)

CAS RN 22014-01-3

IUPAC Name: (2E)-3-(3,5-di-tert-butyl-4-hydroxyphenyl)prop-2-enoic acid
Synonyms: (2E)-3-(3,5-di-tert-butyl-4-hydroxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 276.38
Molecular Formula: C17H24O3
InChi Key: CTYWXRDQWMRIIM-BQYQJAHWSA-N
SMILES: CC(C)(C)C1=CC(\C=C\C(O)=O)=CC(=C1O)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)