missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxycinnamic Acid, TRC

Product Code. 30388166
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388166 5 g
30385905 1 g
30376635 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388166 Supplier Toronto Research Chemicals Supplier No. TRCD6792505G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22014-01-3
Molecular Formula C17H24O3
Molecular Weight (g/mol) 276.37
Synonym 3-[3,5-Bis(1,1-dimethylethyl)-4-hydroxyphenyl]-2-propenoic Acid, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)acrylic Acid, 3-[3,5-Bis(tert-butyl)-4-hydroxyphenyl]acrylic Acid, 3-[4-Hydroxy-3,5-di(tert-butyl)phenyl]-2-propenoic acid, ATACL, NSC 226266
SMILES CC(C)(C)c1cc(\C=C\C(=O)O)cc(c1O)C(C)(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C17H24O3/c1-16(2,3)12-9-11(7-8-14(18)19)10-13(15(12)20)17(4,5)6/h7-10,20H,1-6H3,(H,18,19)/b8-7+
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxycinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.