accessibility menu, dialog, popup

UserName

OHOFFFF

CAS RN 220227-66-7

IUPAC Name: 2-[2-fluoro-5-(trifluoromethyl)phenyl]acetic acid
Synonyms: [2-fluoro-5-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: OIVIPAUIJVTMGM-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(=CC=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results