accessibility menu, dialog, popup

UserName

OOOO

CAS RN 22042-71-3

IUPAC Name: 2-(4-formylphenoxy)acetate
Synonyms: 2-(4-formylphenoxy)acetate
Molecular Weight (g/mol): 179.15
Molecular Formula: C9H7O4
InChi Key: OYNIIKHNXNPSAG-UHFFFAOYSA-M
SMILES: [O-]C(=O)COC1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results