missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenoxyacetic acid, 98%

Product Code. 11461708
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461708 250 g 250g
11441708 10 g 10g
11451708 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461708 Supplier Thermo Scientific Alfa Aesar Supplier No. A13960.30
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22042-71-3 | C9H7O4 | 179.15 g/mol

4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in hot methanol within almost transparency.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22042-71-3
Molecular Formula C9H7O4
Molecular Weight (g/mol) 179.15
MDL Number MFCD00016613
InChI Key OYNIIKHNXNPSAG-UHFFFAOYSA-M
Synonym 4-formylphenoxyacetic acid, 4-formylphenoxy acetic acid, 2-4-formylphenoxy acetic acid, p-formylphenoxyacetic acid, acetic acid, 4-formylphenoxy, acetic acid, 2-4-formylphenoxy, p-formylphenoxy acetic acid, 4-carboxymethyloxy benzaldehyde, 4-formyl phenoxy acetic acid, 4-formylphenoxyactic acid
PubChem CID 89177
IUPAC Name 2-(4-formylphenoxy)acetic acid
SMILES [O-]C(=O)COC1=CC=C(C=O)C=C1

Specifications

Melting Point 193°C to 198°C
Quantity 250 g
Beilstein 779885
Sensitivity Air Sensitive
Solubility Information Soluble in hot methanol within almost transparency.
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 4-Formylphenoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.