accessibility menu, dialog, popup

UserName

OHONNFFF

CAS RN 220462-27-1

IUPAC Name: 4-[3-(trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
Synonyms: 4-[3-(trifluoromethyl)pyrazol-1-yl]benzoic acid
Molecular Weight (g/mol): 256.18
Molecular Formula: C11H7F3N2O2
InChi Key: ZQBFVQMSDDVBSD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)N1C=CC(=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results