missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid, 97%, Thermo Scientific™

Product Code. 10772683
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772683 1 g Amber Glass Bottle 1g
10503222 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772683 Supplier Thermo Scientific Maybridge Supplier No. MO00070DA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 220462-27-1
Molecular Formula C11H7F3N2O2
Molecular Weight (g/mol) 256.184
MDL Number MFCD03086203
InChI Key ZQBFVQMSDDVBSD-UHFFFAOYSA-N
Synonym 4-3-trifluoromethyl-1h-pyrazol-1-yl benzoic acid, 4-3-trifluoromethyl pyrazol-1-yl benzoic acid, maybridge3_004363, 4-3-trifluoromethyl pyrazolyl benzoic acid, 4-3-trifluoromethyl-1-pyrazolyl benzoic acid, 1-4-carboxyphenyl-3-trifluoromethyl-1h-pyrazole, 4-3-trifluoromethyl-1h-pyrazol-1-yl benzoicacid, 4-3-trifluoromethyl-1h-pyrazol-1-yl benzoic acid, benzoic acid,4-3-trifluoromethyl-1h-pyrazol-1-yl
PubChem CID 2780696
IUPAC Name 4-[3-(trifluoromethyl)pyrazol-1-yl]benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)N2C=CC(=N2)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 256.18
Percent Purity 97%
Chemical Name or Material 4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.