accessibility menu, dialog, popup

UserName

OOH

CAS RN 2215-78-3

IUPAC Name: (4-phenoxyphenyl)methanol
Synonyms: (4-phenoxyphenyl)methanol
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: FEOMFFKZOZMBKD-UHFFFAOYSA-N
SMILES: OCC1=CC=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results