missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzyl alcohol, 97%

Product Code. 15475937
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475937 5 g 5g
15485937 250 mg 250mg
15465937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475937 Supplier Thermo Scientific Alfa Aesar Supplier No. H33138.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2215-78-3 | C13H12O2 | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2215-78-3
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD01463970
InChI Key FEOMFFKZOZMBKD-UHFFFAOYSA-N
PubChem CID 826195
IUPAC Name (4-phenoxyphenyl)methanol
SMILES OCC1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Quantity 5 g
Synonym 4-phenoxyphenyl methanol, 4-phenoxybenzyl alcohol, 4-phenoxy-phenyl-methanol, 4-phenoxybenzylalcohol, p-phenoxybenzyl alcohol, benzenemethanol,4-phenoxy, benzenemethanol, 4-phenoxy, 4-phenyloxy phenyl methanol, 4-phenoxyphenyl methan-1-ol
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material 4-Phenoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.