accessibility menu, dialog, popup

UserName

CAS RN 2216-51-5

IUPAC Name: (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Synonyms: menthol L-(-)-menthol U.s.p. menthol (-)-menthol U.S.P. menthol 薄荷脑 D,L-menthol levomentholum L-menthol levomenthol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: NOOLISFMXDJSKH-KXUCPTDWSA-N
SMILES: CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
Molecular Weight (g/mol) 
  • (3)
  • (9)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (4)
Boiling Point 
  • (1)
  • (3)
  • (4)
Physical Form 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (9)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)