accessibility menu, dialog, popup

UserName

HOOO

CAS RN 2243-42-7

IUPAC Name: 2-phenoxybenzoic acid
Synonyms: O-phenoxybenzoic acid
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: PKRSYEPBQPFNRB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1OC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)