missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzoic Acid, TRC

Product Code. 30134366
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134366 10 g 10g
30132390 50 g 50g
30005285 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134366 Supplier Toronto Research Chemicals Supplier No. TRCP39941310G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2243-42-7
Molecular Formula C13 H10 O3
Molecular Weight (g/mol) 214.22
Synonym 2-Phenoxybenzoic Acid, Benzoic acid, 2-phenoxy- (9CI, ACI), 2-Phenoxybenzoic acid (ACI), Benzoic acid, o-phenoxy- (6CI, 7CI, 8CI), 2-(Phenyloxy)benzoic acid, 2-PBA, NSC 39656, o-Phenoxybenzoic acid
IUPAC Name 2-phenoxybenzoic acid
SMILES OC(=O)c1ccccc1Oc2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H10O3/c14-13(15)11-8-4-5-9-12(11)16-10-6-2-1-3-7-10/h1-9H,(H,14,15)
Formula Weight 214.063
Chemical Name or Material 2-Phenoxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.