accessibility menu, dialog, popup

UserName

CAS RN 2265-92-1

FFI

CAS RN 2265-92-1

IUPAC Name: 1,4-difluoro-2-iodobenzene
Synonyms: 1,4-difluoro-2-iodobenzene
Molecular Weight (g/mol): 239.99
Molecular Formula: C6H3F2I
InChi Key: WBYVNDUCUMNZPM-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(I)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results