missing translation for 'onlineSavingsMsg'
Learn More

1,4-Difluoro-2-iodobenzene, 97%, Thermo Scientific Chemicals

Product Code. 11461430
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461430 5 g 5g
11451430 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461430 Supplier Thermo Scientific Alfa Aesar Supplier No. H26890.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2265-92-1
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.99
MDL Number MFCD03094151
InChI Key WBYVNDUCUMNZPM-UHFFFAOYSA-N
PubChem CID 223084
IUPAC Name 1,4-difluoro-2-iodobenzene
SMILES FC1=CC=C(F)C(I)=C1

Specifications

Density 1.998
Boiling Point 178°C to 179°C
Flash Point 85°C (185°F)
Refractive Index 1.563
Quantity 5 g
Sensitivity Light sensitive
Synonym 2,5-difluoroiodobenzene, benzene,1,4-difluoro-2-iodo, 2,5-difluoro-1-iodobenzene, benzene, 1,4-difluoro-2-iodo, pubchem4402, acmc-209fxx, 1,4-bis fluoranyl-2-iodanyl-benzene
Formula Weight 239.99
Percent Purity 97%
Chemical Name or Material 1,4-Difluoro-2-iodobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.