accessibility menu, dialog, popup

UserName

CH3OOOHOH3C(E)

CAS RN 2309-07-1

IUPAC Name: methyl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Synonyms: methyl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: AUJXJFHANFIVKH-GQCTYLIASA-N
SMILES: COC(=O)\C=C\C1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results