accessibility menu, dialog, popup

UserName

ClH2N

CAS RN 2338-18-3

IUPAC Name: 2,3-dihydro-1H-inden-2-amine hydrochloridyl
Synonyms: 2-aminoindane hydrochloridyl
Molecular Weight (g/mol): 168.64
Molecular Formula: C9H11ClN
InChi Key: DLYRFJDJKGGVKT-UHFFFAOYSA-N
SMILES: [Cl].NC1CC2=CC=CC=C2C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results