missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoindan hydrochloride, 98%

Product Code. 11995231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11995231 1 g Glass bottle 1g
11905241 5 g Glass bottle 5g
11915241 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11995231 Supplier Thermo Scientific Acros Supplier No. 103860010
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2338-18-3 | C9H11ClN | 168.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2338-18-3
Molecular Formula C9H11ClN
Molecular Weight (g/mol) 168.64
MDL Number MFCD00012549
InChI Key DLYRFJDJKGGVKT-UHFFFAOYSA-N
Synonym 2-aminoindan hydrochloride, 2-aminoindane hydrochloride, 2,3-dihydro-1h-inden-2-amine hydrochloride, 2-aminoindane hcl, 2-indanamine hydrochloride, 2-indanamine, hydrochloride, 2-indanylamine hydrochloride, su 8629 hydrochloride, indan-2-ylammonium chloride, indan-2-ylamine hydrochloride
PubChem CID 122764
IUPAC Name 2,3-dihydro-1H-inden-2-amine;hydrochloride
SMILES [Cl].NC1CC2=CC=CC=C2C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 246.0°C to 247.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 12,1196
Formula Weight 169.65
Percent Purity 98%
Chemical Name or Material 2-Aminoindan hydrochloride
compliance-icons
Product Identifier
  • 2-Aminoindan hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.