accessibility menu, dialog, popup

UserName

BHOOH

CAS RN 236389-21-2

IUPAC Name: {9,9'-spirobi[fluoren]-2-yl}boronic acid
Synonyms: 9,9'-spirobi[fluoren]-2-ylboronic acid
Molecular Weight (g/mol): 360.22
Molecular Formula: C25H17BO2
InChi Key: WDDLHUWVLROJLA-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)