missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Spirobifluorene-2-boronic acid, 98%

Product Code. 15451779
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451779 5 g 5g
15461779 250 mg 250mg
15441779 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451779 Supplier Thermo Scientific Alfa Aesar Supplier No. H64525.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 236389-21-2 | C25H17BO2 | 360.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 236389-21-2
Molecular Formula C25H17BO2
Molecular Weight (g/mol) 360.22
MDL Number MFCD13248568
InChI Key WDDLHUWVLROJLA-UHFFFAOYSA-N
Synonym 9,9'-spirobi fluoren-2-ylboronic acid, 9,9'-spirobi 9h-fluorene-2-boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, 9,9'-spirobi fluorene-2-ylboronic acid, boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl, 9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, boronic acid, 9,9'-spirobi 9h-fluoren-2-yl, acmc-209g4k, 9,9'-spirobifluorene-2-boronic acid, 9,9'-spirobi fluorene-2-yl boronic acid
PubChem CID 22564369
IUPAC Name 9,9'-spirobi[fluorene]-2-ylboronic acid
SMILES OB(O)C1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Quantity 5 g
Beilstein 10478712
Formula Weight 360.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 9,9'-Spirobifluorene-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.