accessibility menu, dialog, popup

UserName

CAS RN 23873-81-6

IUPAC Name: N-[(1E)-2-nitroso-1,2-diphenylethenyl]hydroxylamine
Synonyms: N-[(1E)-2-nitroso-1,2-diphenylethenyl]hydroxylamine
Molecular Weight (g/mol): 240.26
Molecular Formula: C14H12N2O2
InChi Key: ZXBLMXOUGVRCCH-BUHFOSPRSA-N
SMILES: ON\C(=C(\N=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)