missing translation for 'onlineSavingsMsg'
Learn More

Diphenylglyoxime, 97%, Thermo Scientific™

Product Code. 10667941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10667941 100 g Glass bottle 100g
10168112 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10667941 Supplier Thermo Scientific Acros Supplier No. 408931000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kemiska identifierare

CAS 23873-81-6
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002113
InChI Key JJZONEUCDUQVGR-VCFJNTAESA-N
Synonym diphenylglyoxime, benzil dioxime, ethanedione, diphenyl-, dioxime, e,e, diphenyl glyoxime, beta-benzil dioxime, gamma-benzil dioxime, n-z-2-nitroso-1,2-diphenylethenyl hydroxylamine, unii-ji58npg8m0, unii-pqn86x6991, ji58npg8m0
PubChem CID 5369401
IUPAC Name N-[(Z)-2-nitroso-1,2-diphenylethenyl]hydroxylamine
SMILES O\N=C(\C(=N/O)\C1=CC=CC=C1)/C1=CC=CC=C1

Specifikationer

Melting Point 238.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 15,1081
Solubility Information Solubility in water: insoluble.
Formula Weight 240.26
Percent Purity 97%
Grade Technical
Chemical Name or Material Diphenylglyoxime

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.