missing translation for 'onlineSavingsMsg'
Learn More

Benzil dioxime, 98%

Product Code. 11473454
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473454 100 g 100g
11463454 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473454 Supplier Thermo Scientific Alfa Aesar Supplier No. A10687.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 23873-81-6 | C14H12N2O2 | 240.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23873-81-6
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002113
InChI Key JJZONEUCDUQVGR-VCFJNTAESA-N
Synonym diphenylglyoxime, benzil dioxime, ethanedione, diphenyl-, dioxime, e,e, diphenyl glyoxime, beta-benzil dioxime, gamma-benzil dioxime, n-z-2-nitroso-1,2-diphenylethenyl hydroxylamine, unii-ji58npg8m0, unii-pqn86x6991, ji58npg8m0
PubChem CID 5369401
IUPAC Name N-[(Z)-2-nitroso-1,2-diphenylethenyl]hydroxylamine
SMILES O\N=C(\C(=N/O)\C1=CC=CC=C1)/C1=CC=CC=C1

Specifications

Melting Point ∼238°C (decomposition)
Quantity 100 g
Beilstein 2053615
Merck Index 14,1079
Formula Weight 240.26
Percent Purity 98%
Chemical Name or Material Benzil dioxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.