accessibility menu, dialog, popup

UserName

OOOBr

CAS RN 24050-49-5

IUPAC Name: 7-bromo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Synonyms: 7-bromo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular Weight (g/mol): 277.07
Molecular Formula: C12H5BrO3
InChi Key: LYXFXCSFCWZGNZ-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC=CC3=C2C(=C1)C(=O)OC3=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results