missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1,8-naphthalic anhydride, 97%, Thermo Scientific Chemicals

Product Code. 11348626
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348626 1 g 1g
11358626 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348626 Supplier Thermo Scientific Alfa Aesar Supplier No. L17683.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24050-49-5
Molecular Formula C12H5BrO3
Molecular Weight (g/mol) 277.07
MDL Number MFCD00228162
InChI Key LYXFXCSFCWZGNZ-UHFFFAOYSA-N
Synonym 5-bromobenzo de isochromene-1,3-dione, 3-bromo-1,8-naphthalic anhydride, 5-bromo-1h,3h-benzo de isochromene-1,3-dione, pubchem24244, 5-bromo-benzo de isochromene-1,3-dione, 1h,3h-naphtho 1,8-cd pyran-1,3-dione,5-bromo, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 5-bromo, 11-bromo-3-oxatricyclo 7.3.1.0, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 7-bromo-3-oxatricyclo 7.3.1.0?,__ trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 11-bromo-3-oxatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione
PubChem CID 3907491
IUPAC Name 7-bromo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
SMILES BrC1=CC2=CC=CC3=C2C(=C1)C(=O)OC3=O

Specifications

Melting Point 244°C to 246°C
Quantity 1 g
Beilstein 180001
Sensitivity Moisture sensitive
Formula Weight 277.08
Percent Purity 97%
Chemical Name or Material 3-Bromo-1,8-naphthalic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.