CAS RN 24157-81-1
IUPAC Name:
2,6-bis(propan-2-yl)naphthalene
Synonyms:
DIPN
Molecular Weight (g/mol):
212.34
Molecular Formula:
C16H20
InChi Key:
GWLLTEXUIOFAFE-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC2=CC=C(C=C2C=C1)C(C)C
1
–
2
of
2
results
1
2,6-Diisopropylnaphthalene, TRC
CAS: 24157-81-1 Molecular Formula: C16 H20 Molecular Weight (g/mol): 212.33 Synonym: Naphthalene, 2,6-diisopropyl- (6CI,8CI),2,6-Bis(1-methylethyl)naphthalene,2,6-Diisopropylnaphthalene,2,6-Diisospropylnaphthalene,NSC 166467 IUPAC Name: 2,6-di(propan-2-yl)naphthalene SMILES: CC(C)c1ccc2cc(ccc2c1)C(C)C