missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylnaphthalene, TRC

Product Code. 30138022
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138022 50 g 50g
30134825 5 g 5g
17982945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138022 Supplier Toronto Research Chemicals Supplier No. TRCD45936550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24157-81-1
Molecular Formula C16 H20
Molecular Weight (g/mol) 212.33
Synonym Naphthalene, 2,6-diisopropyl- (6CI,8CI), 2,6-Bis(1-methylethyl)naphthalene, 2,6-Diisopropylnaphthalene, 2,6-Diisospropylnaphthalene, NSC 166467
IUPAC Name 2,6-di(propan-2-yl)naphthalene
SMILES CC(C)c1ccc2cc(ccc2c1)C(C)C

Specifications

Quantity 50 g
InChI Formula InChI=1S/C16H20/c1-11(2)13-5-7-16-10-14(12(3)4)6-8-15(16)9-13/h5-12H,1-4H3
Formula Weight 212.1565
Chemical Name or Material 2,6-Diisopropylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.