accessibility menu, dialog, popup

UserName

NH2OHNH2HO

CAS RN 24171-03-7

IUPAC Name: 2,5-diaminobenzene-1,4-diol
Synonyms: 2,5-diaminobenzene-1,4-diol
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: RLXBOUUYEFOFSW-UHFFFAOYSA-N
SMILES: NC1=CC(O)=C(N)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results