missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminohydroquinone dihydrochloride, 97%

Product Code. 11435087
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435087 0.25 g 0.25g
11425087 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435087 Supplier Thermo Scientific Alfa Aesar Supplier No. B21969.01
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 24171-03-7 | C6H10Cl2N2O2 | 213.058 g/mol

2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

Notes
Air sensitive. It may discolor to black-violet on storage. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24171-03-7
Molecular Formula C6H10Cl2N2O2
Molecular Weight (g/mol) 213.058
MDL Number MFCD00239416
InChI Key NILKAWPWTYPHAH-UHFFFAOYSA-N
Synonym 2,5-diaminobenzene-1,4-diol dihydrochloride, 2,5-diamino-1,4-dihydroxybenzene dihydrochloride, 2,5-diaminohydroquinone dihydrochloride, acmc-20aobi, 1,4-benzenediol, 2,5-diamino-, dihydrochloride, 2,5-diaminobenzene-1,4-diol, chloride, chloride, 1,4-benzenediol,2,5-diamino-,hydrochloride 1:2, 2,5-diaminobenzene-1,4-diol-hydrogen chloride 1/2, 2,5-diaminohydroquinone dihydrochloride, technical at
PubChem CID 16212088
IUPAC Name 2,5-diaminobenzene-1,4-diol;dihydrochloride
SMILES C1=C(C(=CC(=C1O)N)O)N.Cl.Cl

Specifications

Melting Point >300°C
Odor Odorless
Quantity 0.25 g
Sensitivity Air Sensitive
Formula Weight 213.06
Percent Purity 97%
Chemical Name or Material 2,5-Diaminohydroquinone dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.