accessibility menu, dialog, popup

UserName

CAS RN 24484-93-3

CH3OONCl

CAS RN 24484-93-3

IUPAC Name: methyl 4-chloropyridine-2-carboxylate
Synonyms: methyl 4-chloropyridine-2-carboxylate
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: VTENWIPSWAMPKI-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results