accessibility menu, dialog, popup

UserName

FFFOOH(E/Z)

CAS RN 247170-17-8

IUPAC Name: 3-(2,4,5-trifluorophenyl)prop-2-enoic acid
Synonyms: 3-(2,4,5-trifluorophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 202.13
Molecular Formula: C9H5F3O2
InChi Key: KFVQEUPEGKJFCW-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC(F)=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results