accessibility menu, dialog, popup

UserName

CH3H3CCH3H3CNCS

CAS RN 25343-70-8

IUPAC Name: 2-isothiocyanato-1,3-bis(propan-2-yl)benzene
Synonyms: 1,3-diisopropyl-2-isothiocyanatobenzene
Molecular Weight (g/mol): 219.35
Molecular Formula: C13H17NS
InChi Key: HZGOUCYIYIFQHX-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N=C=S
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results