missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylphenyl isothiocyanate, 97%

Product Code. 11363056
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363056 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11363056 Supplier Thermo Scientific Alfa Aesar Supplier No. L10893.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25343-70-8 | C13H17NS | 219.346 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25343-70-8
Molecular Formula C13H17NS
Molecular Weight (g/mol) 219.346
MDL Number MFCD00041341
InChI Key HZGOUCYIYIFQHX-UHFFFAOYSA-N
Synonym 2,6-diisopropylphenyl isothiocyanate, 1,3-diisopropyl-2-isothiocyanatobenzene, 2-isothiocyanato-1,3-bis propan-2-yl benzene, 2,6-diisopropylphenylisothiocyanate, 1,3-diisopropyl-2-isothiocyanato-benzene, 2-isothiocyanato-1,3-di propan-2-yl benzene, acmc-1cue0, 2,6-diisopropylphenyl-isothiocyanate, 2,6-diisopropyl-phenyl isothiocyanate, 2,6-bis methylethyl benzenisothiocyanate
PubChem CID 141194
IUPAC Name 2-isothiocyanato-1,3-di(propan-2-yl)benzene
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N=C=S

Specifications

Boiling Point 139°C to 140°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.583
Quantity 5 g
UN Number UN2922
Beilstein 2416576
Sensitivity Moisture sensitive
Formula Weight 219.35
Percent Purity 97%
Chemical Name or Material 2,6-Diisopropylphenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.