CAS RN 2552-55-8
CAS RN 2552-55-8
IUPAC Name:
2-amino-2-(3-oxo-2,3-dihydro-1,2-oxazol-5-yl)acetic acid
Synonyms:
ibotenic acid
Molecular Weight (g/mol):
158.11
Molecular Formula:
C5H6N2O4
InChi Key:
IRJCBFDCFXCWGO-UHFFFAOYNA-N
SMILES:
NC(C(O)=O)C1=CC(=O)NO1
1
–
2
of
2
results
2
Ibotenic acid, MedChemExpress
MedChemExpress Ibotenic acid has agonist activity at both the N-methyl-D-aspartate (NMDA) and trans-ACPD or metabolotropic quisqualate (Qm) receptor sites.