accessibility menu, dialog, popup

UserName

ONHHCl

CAS RN 25602-68-0

IUPAC Name: 8-azabicyclo[3.2.1]octan-3-one hydrochloride
Synonyms: 8-azabicyclo[3.2.1]octan-3-one hydrochloride
Molecular Weight (g/mol): 161.63
Molecular Formula: C7H12ClNO
InChi Key: MZQWQFWRSDNBPV-UHFFFAOYNA-N
SMILES: Cl.O=C1CC2CCC(C1)N2
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)