missing translation for 'onlineSavingsMsg'
Learn More

Nortropinone hydrochloride, 97%

Product Code. 11956661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11956661 5 g 5g
11946661 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11956661 Supplier Thermo Scientific Acros Supplier No. 436120050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 25602-68-0 | C7H11NO·ClH | 161.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25602-68-0
Molecular Formula C7H11NO·ClH
Molecular Weight (g/mol) 161.63
InChI Key MZQWQFWRSDNBPV-UHFFFAOYSA-N
Synonym nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride
PubChem CID 13091218
IUPAC Name 8-azabicyclo[3.2.1]octan-3-one;hydrochloride
SMILES C1CC2CC(=O)CC1N2.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 194.7°C to 195.8°C
Color Colorless
Elemental Analysis 49.94 to 54.10% C, 8.32 to 9.02% N
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Quantity 5 g
Formula Weight 161.63
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Nortropinone hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Nortropinone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.