missing translation for 'onlineSavingsMsg'
Learn More

Nortropinone hydrochloride, 97%

Product Code. 15431719
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431719 100 g 100g
15411719 5 g 5g
15421719 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431719 Supplier Thermo Scientific Alfa Aesar Supplier No. H64485.22
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25602-68-0 | C7H12ClNO | 161.629 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25602-68-0
Molecular Formula C7H12ClNO
Molecular Weight (g/mol) 161.629
MDL Number MFCD03613582
InChI Key MZQWQFWRSDNBPV-UHFFFAOYSA-N
Synonym nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride
PubChem CID 13091218
IUPAC Name 8-azabicyclo[3.2.1]octan-3-one;hydrochloride
SMILES C1CC2CC(=O)CC1N2.Cl

Specifications

Melting Point 194°C to 196°C
Quantity 100 g
Beilstein 3693524
Sensitivity Hygroscopic
Formula Weight 161.63
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Nortropinone hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.