accessibility menu, dialog, popup

UserName

FFFSH

CAS RN 25697-55-6

IUPAC Name: [3-(trifluoromethyl)phenyl]methanethiol
Synonyms: [3-(trifluoromethyl)phenyl]methanethiol
Molecular Weight (g/mol): 192.2
Molecular Formula: C8H7F3S
InChi Key: CQIQWIMXCPTQPJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(CS)=C1
Grade 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)