missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl mercaptan, tech. 90%

Product Code. 11398005
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398005 1 g 1g
11308015 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398005 Supplier Thermo Scientific Alfa Aesar Supplier No. L08155.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25697-55-6 | C8H7F3S | 192.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25697-55-6
Molecular Formula C8H7F3S
Molecular Weight (g/mol) 192.199
MDL Number MFCD00042435
InChI Key CQIQWIMXCPTQPJ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl mercaptan, 3-trifluoromethyl phenyl methanethiol, 3-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl phenylmethanethiol, 3-trifluoromethyl benzylthiol, m-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl benzylmercaptan, 3-trifluoromethyl phenyl methane-1-thiol, acmc-1cnzu
PubChem CID 141236
IUPAC Name [3-(trifluoromethyl)phenyl]methanethiol
SMILES C1=CC(=CC(=C1)C(F)(F)F)CS

Specifications

Boiling Point 75°C to 77°C (5 mmHg)
Refractive Index 1.49
Quantity 1 g
UN Number UN2810
Beilstein 2520097
Sensitivity Air Sensitive
Formula Weight 192.2
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-(Trifluoromethyl)benzyl mercaptan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.