accessibility menu, dialog, popup

UserName

CAS RN 258278-25-0

H3CCH3CH3CH3NNCH3H3CCH3CH3Cl

CAS RN 258278-25-0

IUPAC Name: 1,3-bis[2,6-bis(propan-2-yl)phenyl]-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium
Synonyms: 1,3-bis(2,6-diisopropylphenyl)-4,5-dihydro-1λ⁵-imidazol-1-ylium
Molecular Weight (g/mol): 391.62
Molecular Formula: C27H39N2
InChi Key: SCEZRJLZOZKPBC-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results