missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%

Product Code. 10210423 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10210423 1 g Glass bottle 1g
10085633 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10210423 Supplier Thermo Scientific Acros Supplier No. 378340010
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 258278-25-0 | C27H39ClN2 | 427.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 258278-25-0
Molecular Formula C27H39ClN2
Molecular Weight (g/mol) 427.07
MDL Number MFCD07369796
InChI Key LWPXTYZKAWSRIP-UHFFFAOYSA-M
Synonym 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium chloride, sipr.hcl, 1,3-bis 2,6-diisopropylphenyl-imidazolidinium-chloride, 1,3-bis 2,6-diisopropylphenyl imidazolidinium chloride, n,n'-2,6-diisopropylphenyl dihydroimidazolium chloride, 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride, 1,3-bis 2,6-diisopropylphenyl imidazolinium chloride?, 1,3-bis-2,6-diisopropylphenyl imidazolidinium chloride, 1,3-bis-2,6-diisopropylphenyl imidazolidiniumchloride, 1,3-bis 2,6-bis isopropyl phenyl imidazolin-3-ium chloride
PubChem CID 2734918
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;chloride
SMILES [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 88% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Formula Weight 427.07
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolidinium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.