accessibility menu, dialog, popup

UserName

BBHOHOOHOHH3CCH3

CAS RN 258865-48-4

IUPAC Name: [7-(dihydroxyboranyl)-9,9-dioctyl-9H-fluoren-2-yl]boronic acid
Synonyms: 7-(dihydroxyboranyl)-9,9-dioctylfluoren-2-ylboronic acid
Molecular Weight (g/mol): 478.29
Molecular Formula: C29H44B2O4
InChi Key: HURJMQMZDPOUOU-UHFFFAOYSA-N
SMILES: CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results